C12H316CE708 Chemcia Scientific, LLC

1-(4-Methoxy-phenyl)-cyclopropanecarbonitrile

  • MDL number MFCD00019212
  • Cas Number : 16728-00-0
  • Molecular Weight : 173.21642

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2120

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2120
Availability details for
Quantity
  • Availability
  • Add to Cart