C12H316CE709 Chemcia Scientific, LLC

1-(5-Amino-2-nitro-phenyl)-ethanone

  • MDL number MFCD00034015
  • Cas Number : 16994-13-1
  • Molecular Weight : 180.16456

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2017

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2017
Availability details for
Quantity
  • Availability
  • Add to Cart