C12H316CE710 Chemcia Scientific, LLC

6-Benzyloxy-benzothiazol-2-ylamine

  • MDL number MFCD09755107
  • Cas Number : 50851-01-9
  • Molecular Weight : 256.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-(benzyloxy)-1,3-benzothiazol-2-amine
Inchi Code 1S/C14H12N2OS/c15-14-16-12-7-6-11(8-13(12)18-14)17-9-10-4-2-1-3-5-10/h1-8H,9H2,(H2,15,16)
Inchi Key DBWISUPGDHAXIT-UHFFFAOYSA-N

Description

Purity 96%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2024

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2024
Availability details for
Quantity
  • Availability
  • Add to Cart