C12H316CE718 Chemcia Scientific, LLC

2-Bromo-4-trifluoromethyl-benzothiazole

  • MDL number MFCD08459020
  • Cas Number : 898748-19-1
  • Molecular Weight : 282.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-4-(trifluoromethyl)-1,3-benzothiazole
Inchi Code 1S/C8H3BrF3NS/c9-7-13-6-4(8(10,11)12)2-1-3-5(6)14-7/h1-3H
Inchi Key ZCDNOJQDHNJLRZ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2171

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2171
Availability details for
Quantity
  • Availability
  • Add to Cart