C12H316CE720 Chemcia Scientific, LLC

6-Chloro-4-hydroxymethyl-nicotinic acid

  • MDL number MFCD24843130
  • Cas Number : 1440526-57-7
  • Molecular Weight : 187.58377

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2096

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2096
Availability details for
Quantity
  • Availability
  • Add to Cart