C12H316CE724 Chemcia Scientific, LLC

2-Bromo-6-tert-butyl-benzothiazole

  • MDL number MFCD08459032
  • Cas Number : 898748-39-5
  • Molecular Weight : 270.19299

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2108

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2108
Availability details for
Quantity
  • Availability
  • Add to Cart