C12H316CE72D Chemcia Scientific, LLC

2,4-Dimethyl-pyrimidine-5-carboxylic acid ethyl ester

  • MDL number MFCD00234561
  • Cas Number : 2226-86-0
  • Molecular Weight : 180.20819

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Reddish Brown Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2133

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2133
Availability details for
Quantity
  • Availability
  • Add to Cart