C12H316CE731 Chemcia Scientific, LLC

1-(2,4-Dimethyl-pyrimidin-5-yl)-ethanone

  • MDL number MFCD00053574
  • Cas Number : 55933-85-2
  • Molecular Weight : 150.1817

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 0~5
Physical Form Reddish Brown Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2122

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2122
Availability details for
Quantity
  • Availability
  • Add to Cart