C12H316CE73D Chemcia Scientific, LLC

2,4-Diiodo-5-trifluoromethyl-phenol

  • MDL number MFCD24843134
  • Cas Number : 1440526-46-4
  • Molecular Weight : 413.90536

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2130

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2130
Availability details for
Quantity
  • Availability
  • Add to Cart