C12H316CE743 Chemcia Scientific, LLC

4-Ethyl-3-nitro-benzoic acid methyl ester

  • MDL number MFCD12172999
  • Cas Number : 51885-79-1
  • Molecular Weight : 209.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 4-ethyl-3-nitrobenzoate
Inchi Code 1S/C10H11NO4/c1-3-7-4-5-8(10(12)15-2)6-9(7)11(13)14/h4-6H,3H2,1-2H3
Inchi Key YAGPCEXMZGXMLN-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2148

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2148
Availability details for
Quantity
  • Availability
  • Add to Cart