C12H316CE744 Chemcia Scientific, LLC

3-Amino-4-ethyl-benzoic acid methyl ester

  • MDL number MFCD17276539
  • Cas Number : 24812-93-9
  • Molecular Weight : 179.22061

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0-5*C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2149

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2149
Availability details for
Quantity
  • Availability
  • Add to Cart