C12H316CE745 Chemcia Scientific, LLC

3-Phenylsydnone

  • MDL number MFCD00205029
  • Cas Number : 120-06-9
  • Molecular Weight : 162.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-phenyl-1,2,3-oxadiazol-3-ium-5-olate
Inchi Code 1S/C8H6N2O2/c11-8-6-10(9-12-8)7-4-2-1-3-5-7/h1-6H
Inchi Key KQEVEDHJIGSXDK-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2134

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2134
Availability details for
Quantity
  • Availability
  • Add to Cart