C12H316CE746 Chemcia Scientific, LLC

2-Ethyl-oxazolo[4,5-b]pyridine

  • MDL number MFCD11110208
  • Cas Number : 52333-88-7
  • Molecular Weight : 148.16576

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2153

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2153
Availability details for
Quantity
  • Availability
  • Add to Cart