C12H316CE747 Chemcia Scientific, LLC

7-Chloro-4H-benzo[1,4]oxazin-3-one

  • MDL number MFCD03019820
  • Cas Number : 27320-99-6
  • Molecular Weight : 183.59552

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Red Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2154

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2154
Availability details for
Quantity
  • Availability
  • Add to Cart