C12H316CE749 Chemcia Scientific, LLC

6-Chloro-benzooxazole

  • MDL number MFCD09027437
  • Cas Number : 227197-72-0
  • Molecular Weight : 153.56903

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2156

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2156
Availability details for
Quantity
  • Availability
  • Add to Cart