C12H316CE750 Chemcia Scientific, LLC

N-(2-Acetyl-phenyl)-acetamide

  • MDL number MFCD00032277
  • Cas Number : 5234-26-4
  • Molecular Weight : 177.20467

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2162

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2162
Availability details for
Quantity
  • Availability
  • Add to Cart