C12H316CE754 Chemcia Scientific, LLC

5-Chloromethyl-isoxazole-4-carboxylic acid ethyl ester

  • MDL number MFCD24843138
  • Cas Number : 80196-63-0
  • Molecular Weight : 189.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-(chloromethyl)isoxazole-4-carboxylate
Inchi Code 1S/C7H8ClNO3/c1-2-11-7(10)5-4-9-12-6(5)3-8/h4H,2-3H2,1H3
Inchi Key ZJJNASDETDEEII-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2166

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2166
Availability details for
Quantity
  • Availability
  • Add to Cart