C12H316CE755 Chemcia Scientific, LLC

2-Iodo-5-nitro-benzoic acid methyl ester

  • MDL number MFCD12172544
  • Cas Number : 112239-00-6
  • Molecular Weight : 307.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-iodo-5-nitrobenzoate
Inchi Code 1S/C8H6INO4/c1-14-8(11)6-4-5(10(12)13)2-3-7(6)9/h2-4H,1H3
Inchi Key INEMMZILZDRYJK-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2168

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2168
Availability details for
Quantity
  • Availability
  • Add to Cart