C12H316CE758 Chemcia Scientific, LLC

2-(4-Benzyloxy-phenyl)-2-methyl-propionitrile

  • MDL number MFCD11036601
  • Cas Number : 167762-83-6
  • Molecular Weight : 251.33114

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2165

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2165
Availability details for
Quantity
  • Availability
  • Add to Cart