C12H316CE75A Chemcia Scientific, LLC

2-Amino-1-(carboxylatomethyl)pyridin-1-ium

  • Cas Number : 126202-06-0
  • Molecular Weight : 152.15401

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Light Pink Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2152

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2152
Availability details for
Quantity
  • Availability
  • Add to Cart