C12H316CE75B Chemcia Scientific, LLC

4-Chloromethyl-2-phenyl-pyrimidine-5-carboxylic acid ethyl ester

  • MDL number MFCD24843139
  • Cas Number : 1440526-40-8
  • Molecular Weight : 276.72491

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2216

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2216
Availability details for
Quantity
  • Availability
  • Add to Cart