C12H316CE761 Chemcia Scientific, LLC

6-Oxo-1,6-dihydro-pyridine-3-carboxylic acid hydrazide

  • MDL number MFCD10698665
  • Cas Number : 134531-63-8
  • Molecular Weight : 153.14159

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2222

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2222
Availability details for
Quantity
  • Availability
  • Add to Cart