C12H316CE76A Chemcia Scientific, LLC

6-Methoxy-1H-benzoimidazole

  • MDL number MFCD09842601
  • Cas Number : 4887-80-3
  • Molecular Weight : 148.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-methoxy-1H-benzimidazole
Inchi Code 1S/C8H8N2O/c1-11-6-2-3-7-8(4-6)10-5-9-7/h2-5H,1H3,(H,9,10)
Inchi Key ILMHAGCURJPNRZ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2265

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2265
Availability details for
Quantity
  • Availability
  • Add to Cart