C12H316CE76E Chemcia Scientific, LLC

3-Acetylamino-4-nitro-thiophene-2-carboxylic acid methyl ester

  • MDL number MFCD02091234
  • Cas Number : 80615-53-8
  • Molecular Weight : 244.22736

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2267

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2267
Availability details for
Quantity
  • Availability
  • Add to Cart