C12H316CE773 Chemcia Scientific, LLC

4-Methoxycarbonylmethyl-piperazine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD02684439
  • Cas Number : 731810-20-1
  • Molecular Weight : 258.32

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 4-(2-methoxy-2-oxoethyl)-1-piperazinecarboxylate
Inchi Code 1S/C12H22N2O4/c1-12(2,3)18-11(16)14-7-5-13(6-8-14)9-10(15)17-4/h5-9H2,1-4H3
Inchi Key NZVUWEZJFULOOK-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2257

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2257
Availability details for
Quantity
  • Availability
  • Add to Cart