C12H316CE774 Chemcia Scientific, LLC

2-Amino-5-bromo-4-methoxy-nicotinic acid

  • MDL number MFCD26131632
  • Cas Number : 1820704-59-3
  • Molecular Weight : 247.04944

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2286

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2286
Availability details for
Quantity
  • Availability
  • Add to Cart