C12H316CE77A Chemcia Scientific, LLC

7,8-Dihydro-5H-pyrano[4,3-c]pyridazin-3-ol

  • MDL number MFCD26131634
  • Cas Number : 1075260-60-4
  • Molecular Weight : 152.15401

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2295

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2295
Availability details for
Quantity
  • Availability
  • Add to Cart