C12H316CE77B Chemcia Scientific, LLC

2-(3-Dimethylamino-1-methoxy-allylidene)-malononitrile

  • MDL number MFCD00975440
  • Cas Number : 95689-38-6
  • Molecular Weight : 177.20752

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. 0~5
Physical Form Red Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2290

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2290
Availability details for
Quantity
  • Availability
  • Add to Cart