C12H316CE77C Chemcia Scientific, LLC

6-Nitroimidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester hydrobromide

  • MDL number MFCD26792545
  • Cas Number : 1803571-36-9
  • Molecular Weight : 316.1129

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Salt Data HBr
Storage temp. 0~5
Physical Form Grey Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2289

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2289
Availability details for
Quantity
  • Availability
  • Add to Cart