C12H316CE789 Chemcia Scientific, LLC

3,4-Dihydro-2H-quinolin-1-ylamine hydrochloride

  • MDL number MFCD28400684
  • Cas Number : 303801-64-1
  • Molecular Weight : 184.67036

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Salt Data HCl
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2419

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2419
Availability details for
Quantity
  • Availability
  • Add to Cart