C12H316CE78A Chemcia Scientific, LLC

2-Dimethylaminomethylene-malononitrile

  • MDL number MFCD00001853
  • Cas Number : 16849-88-0
  • Molecular Weight : 121.14279

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2420

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2420
Availability details for
Quantity
  • Availability
  • Add to Cart