C12H316CE791 Chemcia Scientific, LLC

1-(1,1-Dimethoxy-ethyl)-2-methoxy-benzene

  • MDL number MFCD26131670
  • Cas Number : 72244-69-0
  • Molecular Weight : 196.24837

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2412

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2412
Availability details for
Quantity
  • Availability
  • Add to Cart