C12H316CE794 Chemcia Scientific, LLC

3-(Cyanomethyl-amino)-benzoic acid methyl ester

  • MDL number MFCD16171448
  • Cas Number : 75254-58-9
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-[(cyanomethyl)amino]benzoate
Inchi Code 1S/C10H10N2O2/c1-14-10(13)8-3-2-4-9(7-8)12-6-5-11/h2-4,7,12H,6H2,1H3
Inchi Key RSPWAPRUOTWBQV-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2449

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2449
Availability details for
Quantity
  • Availability
  • Add to Cart