C12H316CE795 Chemcia Scientific, LLC

3-(3,5-Dichloro-2-oxo-2H-pyrazin-1-yl)-benzoic acid methyl ester

  • MDL number MFCD26131677
  • Cas Number : 1820647-18-4
  • Molecular Weight : 299.11516

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2431

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2431
Availability details for
Quantity
  • Availability
  • Add to Cart