C12H316CE797 Chemcia Scientific, LLC

3-(3-Amino-5-chloro-2-oxo-2H-pyrazin-1-yl)-benzoic acid methyl ester

  • MDL number MFCD26131676
  • Cas Number : 1820712-46-6
  • Molecular Weight : 279.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 3-(3-amino-5-chloro-2-oxopyrazin-1(2H)-yl)benzoate
Inchi Code 1S/C12H10ClN3O3/c1-19-12(18)7-3-2-4-8(5-7)16-6-9(13)15-10(14)11(16)17/h2-6H,1H3,(H2,14,15)
Inchi Key OKWNSMVGULDNMH-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yelow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2430

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2430
Availability details for
Quantity
  • Availability
  • Add to Cart