C12H316CE798 Chemcia Scientific, LLC

6-Iodo-[1,2,4]triazolo[1,5-a]pyridin-2-ylamine

  • MDL number MFCD18072711
  • Cas Number : 1245647-65-7
  • Molecular Weight : 260.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-iodo[1,2,4]triazolo[1,5-a]pyridin-2-amine
Inchi Code 1S/C6H5IN4/c7-4-1-2-5-9-6(8)10-11(5)3-4/h1-3H,(H2,8,10)
Inchi Key GGOBBEPQXQCTMC-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2454

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2454
Availability details for
Quantity
  • Availability
  • Add to Cart