C12H316CE79C Chemcia Scientific, LLC

6-Trifluoromethyl-imidazo[1,2-a]pyridine-2-carboxylic acid

  • MDL number MFCD09258865
  • Cas Number : 1018828-69-7
  • Molecular Weight : 230.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-(trifluoromethyl)imidazo[1,2-a]pyridine-2-carboxylic acid
Inchi Code 1S/C9H5F3N2O2/c10-9(11,12)5-1-2-7-13-6(8(15)16)4-14(7)3-5/h1-4H,(H,15,16)
Inchi Key KASXDWUVBQTSFN-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2453

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2453
Availability details for
Quantity
  • Availability
  • Add to Cart