C12H316CE7A4 Chemcia Scientific, LLC

4-Oxo-4,5,6,7-tetrahydro-indazole-1-carboxylic acid tert-butyl ester hydrate

  • MDL number MFCD28400686
  • Cas Number : 955406-75-4
  • Molecular Weight : 254.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl 4-oxo-4,5,6,7-tetrahydro-1H-indazole-1-carboxylate hydrate
Inchi Code 1S/C12H16N2O3.H2O/c1-12(2,3)17-11(16)14-9-5-4-6-10(15)8(9)7-13-14;/h7H,4-6H2,1-3H3;1H2
Inchi Key XMCFGJFTGXDGMS-UHFFFAOYSA-N

Description

Purity 98%
Salt Data H2O
Storage temp. 0~5
Physical Form Tan Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2497

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2497
Availability details for
Quantity
  • Availability
  • Add to Cart