C12H316CE7A8 Chemcia Scientific, LLC

Cyclobutanecarboxylic acid phenylamide

  • MDL number MFCD03784889
  • Cas Number : 15907-95-6
  • Molecular Weight : 175.23236

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2470

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2470
Availability details for
Quantity
  • Availability
  • Add to Cart