C12H316CE7A9 Chemcia Scientific, LLC

2-Bromo-7-fluoro-4-methyl-benzothiazole

  • MDL number MFCD09749155
  • Cas Number : 1019108-45-2
  • Molecular Weight : 246.10215

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Orange Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2471

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2471
Availability details for
Quantity
  • Availability
  • Add to Cart