C12H316CE7AB Chemcia Scientific, LLC

Cyclobutanecarboxylic acid (3-chloro-phenyl)-amide

  • MDL number MFCD03785576
  • Cas Number : 15907-81-0
  • Molecular Weight : 209.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(3-chlorophenyl)cyclobutanecarboxamide
Inchi Code 1S/C11H12ClNO/c12-9-5-2-6-10(7-9)13-11(14)8-3-1-4-8/h2,5-8H,1,3-4H2,(H,13,14)
Inchi Key NKTPEKDVZVAYNU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2473

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2473
Availability details for
Quantity
  • Availability
  • Add to Cart