C12H316CE7AC Chemcia Scientific, LLC

3-Trifluoromethyl-1,4,6,7-tetrahydro-pyrazolo[4,3-c]pyridine-5-carboxylic acid tert-butyl ester

  • MDL number MFCD11044975
  • Cas Number : 1022931-73-2
  • Molecular Weight : 291.27542

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2496

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2496
Availability details for
Quantity
  • Availability
  • Add to Cart