C12H316CE7AF Chemcia Scientific, LLC

Cyclobutanecarboxylic acid (4-benzyloxy-phenyl)-amide

  • MDL number MFCD09431028
  • Cas Number : 925605-98-7
  • Molecular Weight : 281.35763

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2491

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2491
Availability details for
Quantity
  • Availability
  • Add to Cart