C12H316CE7B6 Chemcia Scientific, LLC

3-Amino-6-iodo-pyrazine-2-carboxylic acid

  • MDL number MFCD09909641
  • Cas Number : 875781-48-9
  • Molecular Weight : 265.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-amino-6-iodo-2-pyrazinecarboxylic acid
Inchi Code 1S/C5H4IN3O2/c6-2-1-8-4(7)3(9-2)5(10)11/h1H,(H2,7,8)(H,10,11)
Inchi Key MMXRLDGHYRHOGZ-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2492

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2492
Availability details for
Quantity
  • Availability
  • Add to Cart