C12H316CE7B8 Chemcia Scientific, LLC

3-Nitro-1H-indazole

  • MDL number MFCD02853204
  • Cas Number : 31164-27-9
  • Molecular Weight : 163.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-nitro-1H-indazole
Inchi Code 1S/C7H5N3O2/c11-10(12)7-5-3-1-2-4-6(5)8-9-7/h1-4H,(H,8,9)
Inchi Key OWIRCRREDNEXTA-UHFFFAOYSA-N

Description

Purity 98%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=2482

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2482
Availability details for
Quantity
  • Availability
  • Add to Cart