C12H316CE7BB Chemcia Scientific, LLC

3-Chloro-5-iodo-pyridin-2-ylamine

  • MDL number MFCD09754395
  • Cas Number : 952901-62-1
  • Molecular Weight : 254.45843

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2717

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2717
Availability details for
Quantity
  • Availability
  • Add to Cart