C12H316CE7C3 Chemcia Scientific, LLC

N-(4-Chloro-2-trifluoromethyl-phenyl)-acetamide

  • MDL number MFCD08069858
  • Cas Number : 344-53-6
  • Molecular Weight : 237.61044

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=2742

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=2742
Availability details for
Quantity
  • Availability
  • Add to Cart