C12H316CE7E8 Chemcia Scientific, LLC

[1-(5-Bromo-pyridin-2-yl)-pyrrolidin-3-yl]-dimethylamine

  • MDL number MFCD18254883
  • Cas Number : 690264-82-5
  • Molecular Weight : 270.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-[1-(5-bromo-2-pyridinyl)-3-pyrrolidinyl]-N,N-dimethylamine
Inchi Code 1S/C11H16BrN3/c1-14(2)10-5-6-15(8-10)11-4-3-9(12)7-13-11/h3-4,7,10H,5-6,8H2,1-2H3
Inchi Key XTPJCCKHBQLBST-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=22

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=22
Availability details for
Quantity
  • Availability
  • Add to Cart