C12H316CE7E9 Chemcia Scientific, LLC

1-(4-Benzyl-piperazin-1-yl)-3-hydroxy-2-phenyl-propan-1-one

  • MDL number MFCD19441826
  • Cas Number : 1313712-45-6
  • Molecular Weight : 324.42648

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 92
Storage temp. 0~5
Physical Form Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=23

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=23
Availability details for
Quantity
  • Availability
  • Add to Cart