C12H316CE7EA Chemcia Scientific, LLC

(2S)-8-Methyl-8-aza-bicyclo[3.2.1]octan-3-yl 3-(methylsulfonyloxy)-2-phenylpropanoate

  • MDL number MFCD19441827
  • Cas Number : 1313716-46-9
  • Molecular Weight : 367.46765

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=24

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=24
Availability details for
Quantity
  • Availability
  • Add to Cart